Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PsGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130646.3 | NP_570002.1 | 4 - 5 | 1519 | 53 | |
NM_206610.5 | NP_996333.4 | 4 - 5 | 1519 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |