Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258572.1 | NP_001245501.1 | 4 - 5 | 1361 | 66 | |
NM_001297897.1 | NP_001284826.1 | 4 - 5 | 1035 | 66 | ||
NM_001297898.1 | NP_001284827.1 | 5 - 6 | 943 | 66 | ||
NM_130651.3 | NP_570007.2 | 4 - 5 | 1035 | 66 | ||
GenBank mRNA | AY661807.1 | - | 3 - 4 | 693 | 66 | |
BT022705.1 | - | 4 - 5 | 1038 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |