Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha36-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297899.1 | NP_001284828.1 | 1 - 2 | 388 | 58 | |
NM_130652.4 | NP_570008.1 | 1 - 2 | 232 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |