Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha36-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297899.1 | NP_001284828.1 | 3 - 4 | 870 | 83 | |
NM_130652.4 | NP_570008.1 | 3 - 4 | 714 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |