Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299771.1 | NP_001286700.1 | 1 | 469 | 68 | |
NM_137778.2 | NP_611622.2 | 1 | 469 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |