Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp58Db | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137814.4 | NP_611658.3 | 2 - 3 | 399 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |