Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp58Dc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103939.2 | NP_001097409.1 | 5 - 6 | 2267 | 67 | |
NM_001169780.2 | NP_001163251.1 | 4 - 5 | 2047 | 67 | ||
NM_137815.3 | NP_611659.2 | 5 - 6 | 2309 | 67 | ||
GenBank mRNA | BT023942.1 | - | 4 - 5 | 3026 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |