Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp58Dc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103939.2 | NP_001097409.1 | 6 - 7 | 2716 | 64 | |
NM_001169780.2 | NP_001163251.1 | 5 - 6 | 2496 | 64 | ||
NM_137815.3 | NP_611659.2 | 6 - 7 | 2758 | 64 | ||
GenBank mRNA | BT023942.1 | - | 5 - 6 | 3475 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |