Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mes4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169782.2 | NP_001163253.1 | 1 - 2 | 248 | 101 | |
NM_137825.4 | NP_611669.1 | 1 - 2 | 248 | 101 | ||
GenBank mRNA | BT022745.1 | - | 1 - 2 | 230 | 101 | |
KX531907.1 | - | 1 - 2 | 183 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |