Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299813.1 | NP_001286742.1 | 1 - 2 | 659 | 63 | |
NM_137861.3 | NP_611705.1 | 1 - 2 | 481 | 63 | ||
GenBank mRNA | AY069460.1 | - | 1 - 2 | 472 | 63 | |
KX531768.1 | - | 1 - 2 | 139 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |