Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ReepA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202058.2 | NP_001188987.1 | 6 - 7 | 1297 | 83 | |
NM_001202059.2 | NP_001188988.1 | 5 - 6 | 1337 | 83 | ||
NM_001202061.2 | NP_001188990.2 | 6 - 7 | 1297 | 83 | ||
NM_001299820.1 | NP_001286749.1 | 5 - 6 | 1337 | 83 | ||
NM_137893.4 | NP_611737.2 | 3 - 4 | 513 | 83 | ||
GenBank mRNA | BT021231.1 | - | 3 - 4 | 514 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |