Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299917.1 | NP_001286846.1 | 6 - 7 | 2797 | 102 | |
NM_138090.2 | NP_611934.1 | 6 - 7 | 2636 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |