Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272131.2 | NP_001259060.1 | 2 - 3 | 2164 | 134 | |
NM_001297786.2 | NP_001284715.1 | 2 - 3 | 2164 | 134 | ||
NM_143668.3 | NP_651925.1 | 2 - 3 | 2164 | 134 | ||
GenBank mRNA | AJ277789.1 | - | 2 - 3 | 2118 | 134 | |
BT128699.1 | - | 1 - 2 | 1587 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |