Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298007.1 | NP_001284936.1 | 4 - 5 | 559 | 54 | |
NM_143705.3 | NP_651962.2 | 4 - 5 | 559 | 54 | ||
GenBank mRNA | BT150092.1 | - | 4 - 5 | 533 | 54 | |
BT150473.1 | - | 4 - 5 | 533 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |