Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299259.1 | NP_001286188.1 | 4 - 5 | 1872 | 65 | |
NM_143713.3 | NP_651970.1 | 3 - 4 | 1729 | 65 | ||
GenBank mRNA | BT004908.1 | - | 3 - 4 | 1689 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |