Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143719.3 | NP_651976.1 | 1 | 2427 | 144 | |
GenBank mRNA | BT015271.1 | - | 1 | 2340 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |