Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JYalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110622.3 | NP_001104092.2 | 3 - 4 | 2540 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |