Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274304.1 | NP_001261233.1 | 7 - 8 | 1235 | 64 | |
NM_001274306.1 | NP_001261235.1 | 6 - 7 | 1064 | 64 | ||
NM_001274307.1 | NP_001261236.1 | 5 - 6 | 1043 | 64 | ||
NM_001274308.1 | NP_001261237.1 | 5 - 6 | 1043 | 64 | ||
NM_001274309.1 | NP_001261238.1 | 5 - 6 | 1043 | 64 | ||
NM_167846.3 | NP_728558.5 | 7 - 8 | 1235 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |