Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167856.3 | NP_728573.1 | 4 - 5 | 1626 | 108 | |
NM_167857.4 | NP_728574.1 | 4 - 5 | 1605 | 108 | ||
GenBank mRNA | AF220194.1 | - | 4 - 5 | 1604 | 108 | |
AF231037.1 | - | 4 - 5 | 1537 | 108 | ||
AY051495.1 | - | 4 - 5 | 1600 | 108 | ||
BT001795.1 | - | 4 - 5 | 1552 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |