Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp61F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274324.2 | NP_001261253.1 | 7 - 8 | 3355 | 60 | |
NM_057339.5 | NP_476687.1 | 5 - 6 | 1567 | 60 | ||
NM_057340.4 | NP_476688.1 | 5 - 6 | 1567 | 60 | ||
NM_167874.2 | NP_728600.1 | 5 - 6 | 1528 | 60 | ||
NM_167875.2 | NP_728601.1 | 5 - 6 | 1683 | 60 | ||
GenBank mRNA | AY069713.1 | - | 5 - 6 | 1528 | 60 | |
BT133407.1 | - | 5 - 6 | 1497 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |