Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274330.1 | NP_001261259.1 | 9 - 10 | 2821 | 77 | |
NM_001274331.1 | NP_001261260.1 | 8 - 9 | 2698 | 77 | ||
NM_139361.2 | NP_647618.1 | 8 - 9 | 2698 | 77 | ||
NM_167888.2 | NP_728620.1 | 8 - 9 | 2698 | 77 | ||
NM_167889.2 | NP_728621.1 | 9 - 10 | 2821 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |