Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: promL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259658.3 | NP_001246587.1 | 7 - 8 | 1398 | 67 | |
NM_001274422.1 | NP_001261351.1 | 6 - 7 | 1315 | 67 | ||
NM_001274424.1 | NP_001261353.1 | 6 - 7 | 1517 | 67 | ||
NM_001382193.1 | NP_001368963.1 | 7 - 8 | 1398 | 67 | ||
NM_139513.3 | NP_647770.1 | 6 - 7 | 1239 | 67 | ||
NM_167986.1 | NP_728809.1 | 6 - 7 | 1517 | 67 | ||
NM_167987.4 | NP_728810.1 | 7 - 8 | 1398 | 67 | ||
GenBank mRNA | AF127935.2 | - | 7 - 8 | 1373 | 67 | |
AF197345.1 | - | 6 - 7 | 1195 | 67 | ||
BT031128.1 | - | 6 - 7 | 1197 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |