Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZnT63C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274435.1 | NP_001261364.1 | 5 - 6 | 622 | 75 | |
NM_001274436.1 | NP_001261365.1 | 4 - 5 | 548 | 75 | ||
NM_139544.3 | NP_647801.1 | 4 - 5 | 558 | 75 | ||
NM_168015.2 | NP_728856.1 | 5 - 6 | 622 | 75 | ||
NM_168016.2 | NP_728857.1 | 5 - 6 | 791 | 75 | ||
NM_168017.2 | NP_728858.1 | 5 - 6 | 1043 | 75 | ||
NM_168018.2 | NP_728859.1 | 4 - 5 | 544 | 75 | ||
GenBank mRNA | AY058521.1 | - | 4 - 5 | 516 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |