Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHGPx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168024.2 | NP_728868.1 | 2 - 3 | 768 | 64 | |
NM_168025.2 | NP_728869.1 | 3 - 4 | 634 | 64 | ||
NM_168026.2 | NP_728870.1 | 2 - 3 | 423 | 64 | ||
GenBank mRNA | AY061632.1 | - | 1 - 2 | 1841 | 64 | |
BT011331.1 | - | 2 - 3 | 500 | 64 | ||
BT053727.1 | - | 681 | 64 | |||
BT099660.1 | - | 3 - 4 | 592 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |