Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr64a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168048.2 | NP_728920.1 | 1 - 2 | 190 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |