Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr64b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168049.2 | NP_728921.1 | 2 - 3 | 509 | 99 |
Gene Symbol: Gr64c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259681.1 | NP_001246610.1 | 2 - 3 | 509 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |