Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ens | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104028.2 | NP_001097498.1 | 2 - 3 | 152 | 81 | |
NM_168064.2 | NP_728941.2 | 2 - 3 | 152 | 81 | ||
GenBank mRNA | BT010301.1 | - | 2 - 3 | 110 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |