Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139608.4 | NP_647865.2 | 4 - 5 | 1646 | 81 | |
GenBank mRNA | BT021422.1 | - | 4 - 5 | 1594 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |