Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168111.4 | NP_729054.3 | 3 - 4 | 877 | 81 | |
NM_168112.4 | NP_729055.1 | 3 - 4 | 881 | 81 | ||
NM_168113.3 | NP_729056.1 | 2 - 3 | 2332 | 81 | ||
GenBank mRNA | AY118564.1 | - | 3 - 4 | 877 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |