Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104047.5 | NP_001097517.1 | 11 - 12 | 3341 | 60 | |
NM_001104048.2 | NP_001097518.2 | 13 - 14 | 3001 | 60 | ||
NM_001104049.4 | NP_001097519.2 | 12 - 13 | 2796 | 60 | ||
NM_001274517.3 | NP_001261446.1 | 12 - 13 | 3398 | 60 | ||
NM_001274519.3 | NP_001261448.1 | 12 - 13 | 3371 | 60 | ||
NM_001300018.2 | NP_001286947.1 | 11 - 12 | 4353 | 60 | ||
NM_001347763.1 | NP_001334674.1 | 13 - 14 | 3317 | 60 | ||
NM_001347764.1 | NP_001334675.1 | 13 - 14 | 3353 | 60 | ||
NM_168126.4 | NP_729084.1 | 11 - 12 | 2351 | 60 |
Gene Symbol: CG46320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347801.1 | NP_001334709.1 | 11 - 12 | 3341 | 60 | |
NM_001347802.1 | NP_001334710.1 | 13 - 14 | 3317 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |