Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG32395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168175.3 | NP_729182.3 | 1 - 2 | 966 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |