Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: form3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014571.1 | NP_001014571.1 | 5 - 6 | 1345 | 76 | |
NM_001104057.1 | NP_001097527.1 | 6 - 7 | 1564 | 76 | ||
GenBank mRNA | AB061681.1 | - | 5 - 6 | 1344 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |