Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298034.1 | NP_001284963.1 | 6 - 7 | 1402 | 76 | |
NM_001298035.1 | NP_001284964.1 | 6 - 7 | 1339 | 76 | ||
NM_001298036.1 | NP_001284965.1 | 6 - 7 | 1351 | 76 | ||
NM_132121.3 | NP_572349.1 | 6 - 7 | 1390 | 76 | ||
GenBank mRNA | AY051622.1 | - | 6 - 7 | 1390 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |