Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258638.2 | NP_001245567.1 | 3 - 4 | 1704 | 74 | |
NM_132176.4 | NP_572404.1 | 3 - 4 | 1491 | 74 | ||
GenBank mRNA | AY060985.1 | - | 3 - 4 | 1455 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |