Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir7f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144705.1 | NP_001138177.1 | 1 - 2 | 373 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |