Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298064.1 | NP_001284993.1 | 10 - 11 | 3758 | 82 | |
NM_132193.2 | NP_572421.1 | 10 - 11 | 3758 | 82 | ||
GenBank mRNA | BT150246.1 | - | 10 - 11 | 3525 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |