Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201629.2 | NP_001188558.1 | 2 - 3 | 1078 | 62 | |
NM_001201630.2 | NP_001188559.1 | 2 - 3 | 1078 | 62 | ||
NM_001201631.2 | NP_001188560.1 | 3 - 4 | 879 | 62 | ||
NM_132230.4 | NP_572458.3 | 2 - 3 | 1078 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |