Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cp7Fa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038748.1 | NP_001033837.1 | 3 - 4 | 371 | 87 | |
NM_001382069.1 | NP_001368953.1 | 3 - 4 | 521 | 87 | ||
GenBank mRNA | BT021351.1 | - | 3 - 4 | 371 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |