Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132277.3 | NP_572505.1 | 2 - 3 | 1035 | 66 | |
GenBank mRNA | AB034690.1 | - | 2 - 3 | 1006 | 66 | |
AF181631.2 | - | 2 - 3 | 1007 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |