Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274247.2 | NP_001261176.1 | 3 - 4 | 1590 | 72 | |
NM_001274248.1 | NP_001261177.1 | 1 - 2 | 1246 | 72 | ||
NM_138135.2 | NP_611979.1 | 3 - 4 | 2262 | 72 | ||
GenBank mRNA | AY268106.1 | - | 3 - 4 | 2284 | 72 | |
BT011483.1 | - | 3 - 4 | 2259 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |