Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259601.2 | NP_001246530.1 | 6 - 7 | 1910 | 70 | |
NM_001274262.1 | NP_001261191.1 | 6 - 7 | 1913 | 70 | ||
NM_001274263.1 | NP_001261192.1 | 6 - 7 | 1910 | 70 | ||
NM_001274264.1 | NP_001261193.1 | 6 - 7 | 1913 | 70 | ||
NM_138175.4 | NP_612019.2 | 6 - 7 | 1913 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |