Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tudor-SN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274266.1 | NP_001261195.1 | 3 - 4 | 2542 | 80 | |
NM_138177.3 | NP_612021.1 | 3 - 4 | 2542 | 80 | ||
GenBank mRNA | BT003270.1 | - | 3 - 4 | 2516 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |