Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274273.1 | NP_001261202.1 | 6 - 7 | 6191 | 101 | |
NM_138180.4 | NP_612024.4 | 6 - 7 | 6194 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |