Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274273.1 | NP_001261202.1 | 7 - 8 | 6705 | 88 | |
NM_138180.4 | NP_612024.4 | 7 - 8 | 6708 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |