Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138236.2 | NP_612080.1 | 6 - 7 | 1454 | 78 | |
GenBank mRNA | BT001418.1 | - | 6 - 7 | 1430 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |