Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FucTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138263.2 | NP_612107.1 | 1 - 2 | 1156 | 74 | |
GenBank mRNA | AF441265.1 | - | 1 - 2 | 925 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |