Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139365.3 | NP_647622.1 | 2 - 3 | 844 | 82 | |
GenBank mRNA | AY058458.1 | - | 2 - 3 | 844 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |