Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr62Bc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274364.1 | NP_001261293.1 | 2 - 3 | 228 | 84 | |
NM_139411.2 | NP_647668.1 | 2 - 3 | 228 | 84 | ||
GenBank mRNA | AY061158.1 | - | 2 - 3 | 205 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |