Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scramb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139504.3 | NP_647761.1 | 2 - 3 | 671 | 64 | |
GenBank mRNA | AY060271.1 | - | 2 - 3 | 745 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |