Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139853.2 | NP_648110.2 | 1 - 2 | 202 | 62 | |
NM_168207.3 | NP_729244.2 | 1 - 2 | 349 | 62 | ||
NM_168209.4 | NP_729246.2 | 1 - 2 | 202 | 62 | ||
GenBank mRNA | BT023952.1 | - | 1 - 2 | 258 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |